COM448685822 Combi-Blocks, Inc.

Methyl 4-(4-aminophenyl)-2-fluorobenzoate

  • MDL number MFCD20231505
  • Cas Number : 1334500-05-8
  • Molecular Weight : 245.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4467

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4467
Availability details for
Quantity
  • Availability
  • Add to Cart