COM448685826 Combi-Blocks, Inc.

3-[3-(Benzyloxy)phenyl]phenylacetic acid

  • MDL number MFCD20231507
  • Cas Number : 1013405-79-2
  • Molecular Weight : 318.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4471

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4471
Availability details for
Quantity
  • Availability
  • Add to Cart