COM448685827 Combi-Blocks, Inc.

{3-[3-(trifluoromethyl)phenyl]phenyl}acetic acid

  • MDL number MFCD05980213
  • Cas Number : 1224742-15-7
  • Molecular Weight : 280.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4472

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4472
Availability details for
Quantity
  • Availability
  • Add to Cart