COM448685833 Combi-Blocks, Inc.

3-(2-Chloro-4-methoxyphenyl)phenylacetic acid

  • MDL number MFCD20441952
  • Cas Number : 1345472-32-3
  • Molecular Weight : 276.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4478

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4478
Availability details for
Quantity
  • Availability
  • Add to Cart