COM448685838 Combi-Blocks, Inc.

3-Fluorofluoren-9-one

  • MDL number MFCD00034619
  • Cas Number : 1514-15-4
  • Molecular Weight : 198.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4486

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4486
Availability details for
Quantity
  • Availability
  • Add to Cart