COM448685843 Combi-Blocks, Inc.

4-[2-(Benzyloxy)phenyl]thiophene-2-carboxylic acid

  • MDL number MFCD21333113
  • Cas Number : 1355247-35-6
  • Molecular Weight : 310.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4492

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4492
Availability details for
Quantity
  • Availability
  • Add to Cart