COM448685845 Combi-Blocks, Inc.

Methyl 2-chloro-5-phenylbenzoate

  • MDL number MFCD20441956
  • Cas Number : 886969-94-4
  • Molecular Weight : 246.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4494

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4494
Availability details for
Quantity
  • Availability
  • Add to Cart