COM448685847 Combi-Blocks, Inc.

Methyl 5-(4-methanesulfonylphenyl)pyridine-3-carboxylate

  • MDL number MFCD06802745
  • Cas Number : 893740-18-6
  • Molecular Weight : 291.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4497

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4497
Availability details for
Quantity
  • Availability
  • Add to Cart