COM448685848 Combi-Blocks, Inc.

5-(4-Chlorophenyl)-2-methoxypyridine

  • MDL number MFCD20491458
  • Cas Number : 1198416-70-4
  • Molecular Weight : 219.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4498

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4498
Availability details for
Quantity
  • Availability
  • Add to Cart