COM448685849 Combi-Blocks, Inc.

2-(4-Nitrophenyl)pyridine-3-carbonitrile

  • MDL number MFCD20491459
  • Cas Number : 1352318-15-0
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4499

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4499
Availability details for
Quantity
  • Availability
  • Add to Cart