COM448685850 Combi-Blocks, Inc.

3-(4-Nitrophenyl)pyridine-2-carbonitrile

  • MDL number MFCD20491460
  • Cas Number : 1352318-13-8
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4500

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4500
Availability details for
Quantity
  • Availability
  • Add to Cart