COM448685858 Combi-Blocks, Inc.

5-(2,4-Difluorophenyl)-2-methoxypyrimidine

  • MDL number MFCD20529533
  • Cas Number : 1352318-16-1
  • Molecular Weight : 222.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4522

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4522
Availability details for
Quantity
  • Availability
  • Add to Cart