COM448685859 Combi-Blocks, Inc.

Methyl 5-(4-methylphenyl)pyridine-2-carboxylate

  • MDL number MFCD21333114
  • Cas Number : 1355246-89-7
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4523

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4523
Availability details for
Quantity
  • Availability
  • Add to Cart