COM448685860 Combi-Blocks, Inc.

2-Fluoro-4-[4-(methylsulfanyl)phenyl]benzaldehyde

  • MDL number MFCD08701420
  • Cas Number : 893637-41-7
  • Molecular Weight : 246.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4525

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4525
Availability details for
Quantity
  • Availability
  • Add to Cart