COM448685861 Combi-Blocks, Inc.

2-Methoxy-5-(3-nitrophenyl)pyridine

  • MDL number MFCD21333115
  • Cas Number : 939428-15-6
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4528

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4528
Availability details for
Quantity
  • Availability
  • Add to Cart