COM448685862 Combi-Blocks, Inc.

4-(3-Methoxypyridin-2-yl)benzoic acid

  • MDL number MFCD16877924
  • Cas Number : 849757-80-8
  • Molecular Weight : 229.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4529

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4529
Availability details for
Quantity
  • Availability
  • Add to Cart