COM448685866 Combi-Blocks, Inc.

3-(4-Fluorophenyl)pyridine-2-carbonitrile

  • MDL number MFCD14698978
  • Cas Number : 1214340-31-4
  • Molecular Weight : 198.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4533

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4533
Availability details for
Quantity
  • Availability
  • Add to Cart