COM448685867 Combi-Blocks, Inc.

2-(4-Fluorophenyl)pyridine-3-carbonitrile

  • MDL number MFCD14698966
  • Cas Number : 1214335-17-7
  • Molecular Weight : 198.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4534

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4534
Availability details for
Quantity
  • Availability
  • Add to Cart