COM448685871 Combi-Blocks, Inc.

Methyl 2-(4-hydroxyphenyl)benzoate

  • MDL number MFCD21609727
  • Cas Number : 170304-68-4
  • Molecular Weight : 228.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4538

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4538
Availability details for
Quantity
  • Availability
  • Add to Cart