COM448685874 Combi-Blocks, Inc.

4-(3,5-Dichlorophenyl)-2-fluorobenzaldehyde

  • MDL number MFCD22192557
  • Cas Number : 1361871-90-0
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4541

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4541
Availability details for
Quantity
  • Availability
  • Add to Cart