COM448685875 Combi-Blocks, Inc.

4-Fluoro-5'-nitrobiphenyl-3,3'-dicarboxylic acid

  • MDL number MFCD22192558
  • Cas Number : 1373232-53-1
  • Molecular Weight : 305.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4542

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4542
Availability details for
Quantity
  • Availability
  • Add to Cart