COM448685877 Combi-Blocks, Inc.

3-Fluoro-5-phenylbenzonitrile

  • MDL number MFCD14701726
  • Cas Number : 1214352-57-4
  • Molecular Weight : 197.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4546

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4546
Availability details for
Quantity
  • Availability
  • Add to Cart