COM448685881 Combi-Blocks, Inc.

2-Fluoro-5-(4-formyl-3-nitrophenyl)benzoic acid

  • MDL number MFCD22549354
  • Cas Number : 1403483-64-6
  • Molecular Weight : 289.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4551

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4551
Availability details for
Quantity
  • Availability
  • Add to Cart