COM448685887 Combi-Blocks, Inc.

3-(2-Methylphenyl)pyridine-2-carbonitrile

  • MDL number MFCD22586705
  • Cas Number : 1415819-89-4
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4557

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4557
Availability details for
Quantity
  • Availability
  • Add to Cart