COM448685890 Combi-Blocks, Inc.

3-(2-Hydroxyphenyl)pyridine-2-carbonitrile

  • MDL number MFCD22689142
  • Cas Number : 1420800-31-2
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4660

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4660
Availability details for
Quantity
  • Availability
  • Add to Cart