COM448685893 Combi-Blocks, Inc.

3-[4-(Hydroxymethyl)phenyl]pyridine-2-carbonitrile

  • MDL number MFCD23135975
  • Cas Number : 1423037-18-6
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4663

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4663
Availability details for
Quantity
  • Availability
  • Add to Cart