COM448685899 Combi-Blocks, Inc.

2-Fluoro-4-phenylbenzaldehyde

  • MDL number MFCD02261762
  • Cas Number : 183436-80-8
  • Molecular Weight : 200.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4671

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4671
Availability details for
Quantity
  • Availability
  • Add to Cart