COM448685905 Combi-Blocks, Inc.

Methyl 4-(2,5-dimethoxyphenyl)benzoate

  • MDL number MFCD24448855
  • Cas Number : 1437794-61-0
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4680

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4680
Availability details for
Quantity
  • Availability
  • Add to Cart