COM448685906 Combi-Blocks, Inc.

3-(4-Cyanophenyl)-5-methoxybenzonitrile

  • MDL number MFCD25121774
  • Cas Number : 1445322-49-5
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4681

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4681
Availability details for
Quantity
  • Availability
  • Add to Cart