COM448685912 Combi-Blocks, Inc.

Methyl 2-amino-5-(4-fluorophenyl)pyridine-4-carboxylate

  • MDL number MFCD14701169
  • Cas Number : 1214354-57-0
  • Molecular Weight : 246.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4690

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4690
Availability details for
Quantity
  • Availability
  • Add to Cart