COM448685915 Combi-Blocks, Inc.

Methyl 5-(2,4-difluorophenyl)pyridine-3-carboxylate

  • MDL number MFCD19704379
  • Cas Number : 1346691-66-4
  • Molecular Weight : 249.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4693

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4693
Availability details for
Quantity
  • Availability
  • Add to Cart