COM448685918 Combi-Blocks, Inc.

Methyl 6-(2,4-difluorophenyl)pyridine-2-carboxylate

  • MDL number MFCD27981348
  • Cas Number : 1820619-37-1
  • Molecular Weight : 249.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4698

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4698
Availability details for
Quantity
  • Availability
  • Add to Cart