COM448685919 Combi-Blocks, Inc.

4-[4-(1H-Pyrazol-4-yl)phenyl]-1H-pyrazole

  • MDL number MFCD23704797
  • Cas Number : 1036248-62-0
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4699

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4699
Availability details for
Quantity
  • Availability
  • Add to Cart