COM448685920 Combi-Blocks, Inc.

3-(2-Methoxypyridin-4-yl)benzoic acid

  • MDL number MFCD25242237
  • Cas Number : 1554504-76-5
  • Molecular Weight : 229.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4700

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4700
Availability details for
Quantity
  • Availability
  • Add to Cart