COM448685921 Combi-Blocks, Inc.

(2-Hydroxy-4-phenylphenyl)thiourea

  • MDL number MFCD28096662
  • Cas Number : 1820608-16-9
  • Molecular Weight : 244.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4720

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4720
Availability details for
Quantity
  • Availability
  • Add to Cart