COM448685927 Combi-Blocks, Inc.

4-[3-(Benzyloxy)-5-fluorophenyl]-2-methoxybenzoic acid

  • MDL number MFCD28166418
  • Cas Number : 1820618-85-6
  • Molecular Weight : 352.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4729

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4729
Availability details for
Quantity
  • Availability
  • Add to Cart