COM448685931 Combi-Blocks, Inc.

Cbz-l-valine methyl ester

  • MDL number MFCD00672538
  • Cas Number : 24210-19-3
  • Molecular Weight : 265.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0005

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0005
Availability details for
Quantity
  • Availability
  • Add to Cart