COM448685935 Combi-Blocks, Inc.

(2E)-3-(2-Bromo-4-fluorophenyl)-2-propenoic acid, methyl ester

  • MDL number MFCD22124745
  • Cas Number : 404575-30-0
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0029

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0029
Availability details for
Quantity
  • Availability
  • Add to Cart