COM448685941 Combi-Blocks, Inc.

1,4-dimethyl (2R,3R)-2,3-bis[(4-methylphenyl)carbonyloxy]butanedioate

  • MDL number MFCD27578184
  • Cas Number : 1773493-87-0
  • Molecular Weight : 414.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0059

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0059
Availability details for
Quantity
  • Availability
  • Add to Cart