COM448685947 Combi-Blocks, Inc.

Dimethyl 1-methyl-1H-imidazole-4,5-dicarboxylate

  • MDL number MFCD00553098
  • Cas Number : 42545-22-2
  • Molecular Weight : 198.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0124

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0124
Availability details for
Quantity
  • Availability
  • Add to Cart