COM448685948 Combi-Blocks, Inc.

Methyl (2S)-2-amino-5-(carbamoylamino)pentanoate

  • MDL number MFCD27578185
  • Cas Number : 742655-88-5
  • Molecular Weight : 189.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0130

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0130
Availability details for
Quantity
  • Availability
  • Add to Cart