COM448685950 Combi-Blocks, Inc.

N-Alpha-carbobenzoxy-d-alanine methyl ester

  • MDL number MFCD00136584
  • Cas Number : 68223-02-9
  • Molecular Weight : 237.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0149

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0149
Availability details for
Quantity
  • Availability
  • Add to Cart