COM448685956 Combi-Blocks, Inc.

Methyl 2-(2-bromo-4-fluorophenyl)acetate

  • MDL number MFCD20527142
  • Cas Number : 949168-34-7
  • Molecular Weight : 247.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0170

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0170
Availability details for
Quantity
  • Availability
  • Add to Cart