COM448685959 Combi-Blocks, Inc.

Methyl N2-(tert-butoxycarbonyl)-L-asparaginate

  • MDL number MFCD08275816
  • Cas Number : 124842-28-0
  • Molecular Weight : 246.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0179

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0179
Availability details for
Quantity
  • Availability
  • Add to Cart