COM448685960 Combi-Blocks, Inc.

Methyl 3-(2-chlorophenyl)propanoate

  • MDL number MFCD16620959
  • Cas Number : 74124-86-0
  • Molecular Weight : 198.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0183

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0183
Availability details for
Quantity
  • Availability
  • Add to Cart