COM448685963 Combi-Blocks, Inc.

methyl 3-(N',N'-dimethylhydrazinecarbonyl)propanoate

  • MDL number MFCD27578190
  • Cas Number : 28402-64-4
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0190

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0190
Availability details for
Quantity
  • Availability
  • Add to Cart