COM448685966 Combi-Blocks, Inc.

(2E)-3-(2,5-Dimethoxyphenyl)-2-propenoic acid, methyl ester

  • MDL number MFCD18446121
  • Cas Number : 116406-20-3
  • Molecular Weight : 222.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0195

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0195
Availability details for
Quantity
  • Availability
  • Add to Cart