COM448685967 Combi-Blocks, Inc.

Methyl 3-(3',4'-dimethoxyphenyl)propanoate

  • MDL number MFCD07369514
  • Cas Number : 27798-73-8
  • Molecular Weight : 224.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0197
Availability details for
Quantity
  • Availability
  • Add to Cart