COM448685972 Combi-Blocks, Inc.

4-Amino-3-(4-chloro-phenyl)-butyric acid methyl ester

  • MDL number MFCD00824612
  • Cas Number : 446053-47-0
  • Molecular Weight : 227.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0204

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0204
Availability details for
Quantity
  • Availability
  • Add to Cart