COM448685974 Combi-Blocks, Inc.

Methyl 2,3,5,6-tetrafluorobenzoate

  • MDL number MFCD06203743
  • Cas Number : 4707-12-4
  • Molecular Weight : 208.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0207

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-0207
Availability details for
Quantity
  • Availability
  • Add to Cart